ENA424045463 Enamine

1-(5-chloropyrazin-2-yl)ethan-1-amine

  • MDL number MFCD20726757
  • Cas Number : 1234014-51-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. 0

Safety Information

signalword Danger
msds https://www.enaminestore.com/catalog/EN300-174550
pictograms GHS05,GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart