ENA424045533 Enamine

5-fluoro-1,3-benzothiazole-2-carbaldehyde

  • MDL number MFCD23788131
  • Cas Number : 139425-46-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. 0

Safety Information

msds https://www.enaminestore.com/catalog/EN300-174762
Availability details for
Quantity
  • Availability
  • Add to Cart