ENA424045534 Enamine

2-(ethylamino)acetamide hydrochloride

  • MDL number MFCD00154700
  • Cas Number : 6913-24-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Normal

Safety Information

msds https://www.enaminestore.com/catalog/EN300-174763
Availability details for
Quantity
  • Availability
  • Add to Cart