ENA424045540 Enamine

2-(cyclopentyloxy)-4-methoxybenzoic acid

  • MDL number MFCD13475649
  • Cas Number : 1292690-31-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Normal

Safety Information

msds https://www.enaminestore.com/catalog/EN300-174780
Availability details for
Quantity
  • Availability
  • Add to Cart