ENA424045578 Enamine

5-methanesulfonylquinolin-8-amine

  • MDL number MFCD12173054
  • Cas Number : 1154236-71-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Normal

Safety Information

msds https://www.enaminestore.com/catalog/EN300-174904
Availability details for
Quantity
  • Availability
  • Add to Cart