ENA424045649 Enamine

3-benzylpyrrolidine-2,5-dione

  • MDL number MFCD01683244
  • Cas Number : 66195-04-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Normal

Safety Information

msds https://www.enaminestore.com/catalog/EN300-175169
Availability details for
Quantity
  • Availability
  • Add to Cart