ENA424045675 Enamine

2-chloro-N-(propan-2-yl)-5-(trifluoromethyl)benzene-1-sulfonamide

  • MDL number MFCD20538853
  • Cas Number : 1378306-70-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Normal

Safety Information

msds https://www.enaminestore.com/catalog/EN300-175269
Availability details for
Quantity
  • Availability
  • Add to Cart