ENA424045758 Enamine

(3-bromo-4-methylphenyl)methanamine

  • MDL number MFCD18392011
  • Cas Number : 1177558-32-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. 0

Safety Information

msds https://www.enaminestore.com/catalog/EN300-175560
Availability details for
Quantity
  • Availability
  • Add to Cart