ENA424045805 Enamine

7-(tetramethyl-1,3,2-dioxaborolan-2-yl)-2,3,4,5-tetrahydro-1H-2-benzazepin-1-one

  • MDL number MFCD18383839
  • Cas Number : 1215007-07-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Normal

Safety Information

msds https://www.enaminestore.com/catalog/EN300-175728
Availability details for
Quantity
  • Availability
  • Add to Cart