ENA424045900 Enamine

3-(1-methyl-1H-pyrazol-4-yl)-1$l^{6},4-thiomorpholine-1,1-dione dihydrochloride

  • MDL number MFCD28012435
  • Cas Number : 1788530-14-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Normal

Safety Information

msds https://www.enaminestore.com/catalog/EN300-176133
Availability details for
Quantity
  • Availability
  • Add to Cart