ENA424045917 Enamine

2-(phenoxymethyl)-1,4-dihydropyrimidin-4-one hydrochloride

  • MDL number MFCD28012452
  • Cas Number : 1790140-56-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-176486

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-176486
Availability details for
Quantity
  • Availability
  • Add to Cart