ENA424045919 Enamine

1-{1-methyl-1H-pyrazolo[3,4-d]pyrimidin-4-yl}piperidine-2-carboxylic acid hydrochloride

  • MDL number MFCD28012454
  • Cas Number : 1803570-61-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-176502

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-176502
Availability details for
Quantity
  • Availability
  • Add to Cart