ENA424045920 Enamine

ethyl 2-(pyrrolidin-2-yl)-1,3-thiazole-4-carboxylate hydrobromide

  • MDL number MFCD28012455
  • Cas Number : 1798725-69-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HBr
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-176626

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-176626
Availability details for
Quantity
  • Availability
  • Add to Cart