ENA424045921 Enamine

N-[(1H-1,2,3-triazol-5-yl)methyl]aniline

  • MDL number MFCD28155300
  • Cas Number : 165397-77-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-176854

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-176854
Availability details for
Quantity
  • Availability
  • Add to Cart