ENA424045929 Enamine

[4-(prop-1-yn-1-yl)phenyl]methanol

  • MDL number MFCD28012465
  • Cas Number : 1360803-46-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-177518

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-177518
Availability details for
Quantity
  • Availability
  • Add to Cart