ENA424045930 Enamine

5-[(2R,4S)-4-methoxypyrrolidin-2-yl]-3-methyl-1,2,4-oxadiazole hydrochloride

  • MDL number MFCD28012466
  • Cas Number : 1808338-33-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-177561

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-177561
Availability details for
Quantity
  • Availability
  • Add to Cart