ENA424045937 Enamine

4-(4H-1,2,4-triazol-4-yl)aniline hydrochloride

  • MDL number MFCD28012474
  • Cas Number : 1803598-03-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-69100

Safety Information

msds https://www.enaminestore.com/catalog/EN300-69100
Availability details for
Quantity
  • Availability
  • Add to Cart