ENA424045973 Enamine

1-(2-chlorophenyl)cyclobutan-1-ol

  • MDL number MFCD21788632
  • Cas Number : 1511366-61-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Normal

Safety Information

msds https://www.enaminestore.com/catalog/EN300-176129
Availability details for
Quantity
  • Availability
  • Add to Cart