ENA424045976 Enamine

tert-butyl N-(2-chloroethyl)-N-methylcarbamate

  • MDL number MFCD20624610
  • Cas Number : 220074-38-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20
Shipping Temperature Dry Ice
Product Link https://www.enaminestore.com/catalog/EN300-175871

Safety Information

signalword Danger
pcodes P264,P270,P280,P301+P312,P302+P352,P305+P351+P338,P310,P330,P332+P313,P362,P501
hcodes Xn
MSDS https://www.enaminestore.com/catalog/EN300-175871
pictograms GHS05,GHS07
hstatements H302,H315,H318
Availability details for
Quantity
  • Availability
  • Add to Cart