ENA424046048 Enamine

(4S)-4-amino-5-fluoropentanoic acid hydrochloride

  • MDL number MFCD28012451
  • Cas Number : 72522-16-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. 0

Safety Information

msds https://www.enaminestore.com/catalog/EN300-176471
Availability details for
Quantity
  • Availability
  • Add to Cart