ENA424046052 Enamine

2-(2-{[(tert-butoxy)carbonyl]amino}pyridin-3-yl)acetic acid

  • MDL number MFCD11848315
  • Cas Number : 1260880-17-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-176529

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-176529
Availability details for
Quantity
  • Availability
  • Add to Cart