ENA424046060 Enamine

1-(cyclopropylmethyl)-1H-indole-2-carboxylic acid

  • MDL number MFCD20185192
  • Cas Number : 1483284-21-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-176612

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-176612
Availability details for
Quantity
  • Availability
  • Add to Cart