ENA424046062 Enamine

ethyl 2-(pyrrolidin-2-yl)-1,3-thiazole-4-carboxylate

  • MDL number MFCD21294932
  • Cas Number : 1190775-48-4
  • Molecular Weight : 226.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-176627

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-176627
Availability details for
Quantity
  • Availability
  • Add to Cart