ENA424046081 Enamine

1-(2-chlorophenyl)-5-(propan-2-yl)-1H-pyrazole-4-carboxylic acid

  • MDL number MFCD12069391
  • Cas Number : 1002249-52-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-177074

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-177074
Availability details for
Quantity
  • Availability
  • Add to Cart