ENA424046087 Enamine

1-(2-methylphenyl)-5-(propan-2-yl)-1H-pyrazol-4-amine

  • MDL number MFCD21773837
  • Cas Number : 1245007-64-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-177205

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-177205
Availability details for
Quantity
  • Availability
  • Add to Cart