ENA424046091 Enamine

N-(pyridin-2-yl)prop-2-enamide

  • MDL number MFCD00596317
  • Cas Number : 40000-69-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-177239

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-177239
Availability details for
Quantity
  • Availability
  • Add to Cart