ENA424046093 Enamine

methyl 2-bromo-3-{[(tert-butoxy)carbonyl]amino}propanoate

  • MDL number MFCD24470484
  • Cas Number : 1207950-16-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. -20

Safety Information

msds https://www.enaminestore.com/catalog/EN300-177244
Availability details for
Quantity
  • Availability
  • Add to Cart