ENA424046094 Enamine

tert-butyl 5-methyl-2,2-dioxo-1,2lambda6,3-oxathiazolidine-3-carboxylate

  • MDL number MFCD24532912
  • Cas Number : 1365481-13-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-177250

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-177250
Availability details for
Quantity
  • Availability
  • Add to Cart