ENA424046097 Enamine

N-(2-methoxyethyl)-N-methyl-4-(methylamino)benzene-1-sulfonamide

  • MDL number MFCD13488885
  • Cas Number : 1303894-24-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-177261

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-177261
Availability details for
Quantity
  • Availability
  • Add to Cart