ENA424046099 Enamine

1-{bicyclo[2.2.1]heptan-2-yl}methanamine hydrochloride

  • MDL number MFCD00034956
  • Cas Number : 4040-59-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 210-215
Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-177265

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-177265
Availability details for
Quantity
  • Availability
  • Add to Cart