ENA424046110 Enamine

(9H-fluoren-9-yl)methyl N-(2-hydroxycyclohexyl)carbamate

  • MDL number MFCD18328487
  • Cas Number : 1342698-64-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-177354

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-177354
Availability details for
Quantity
  • Availability
  • Add to Cart