ENA424046115 Enamine

1-(5-bromo-1-ethyl-1H-1,3-benzodiazol-2-yl)methanamine

  • MDL number MFCD12150191
  • Cas Number : 1183776-28-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-177367

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-177367
Availability details for
Quantity
  • Availability
  • Add to Cart