ENA424046119 Enamine

2-(2-methylpropyl)benzoic acid

  • MDL number MFCD19598685
  • Cas Number : 100058-55-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-177374

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-177374
Availability details for
Quantity
  • Availability
  • Add to Cart