ENA424046120 Enamine

3-(2,2,3,3-tetrafluoropropoxy)propanoic acid

  • MDL number MFCD10693822
  • Cas Number : 1152591-07-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-177379

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-177379
Availability details for
Quantity
  • Availability
  • Add to Cart