ENA424046121 Enamine

2,6-difluoro-4-methoxybenzene-1-sulfonamide

  • MDL number MFCD18394894
  • Cas Number : 1261738-62-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-177380

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-177380
Availability details for
Quantity
  • Availability
  • Add to Cart