ENA424046122 Enamine

2-(methoxymethyl)-2-(2-methylpropyl)pyrrolidine

  • MDL number MFCD21955365
  • Cas Number : 1508970-30-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-177381

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-177381
Availability details for
Quantity
  • Availability
  • Add to Cart