ENA424046127 Enamine

3-ethylimidazolidin-4-one

  • MDL number MFCD26027198
  • Cas Number : 1541613-08-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-177398

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-177398
Availability details for
Quantity
  • Availability
  • Add to Cart