ENA424046131 Enamine

ethyl 5-chloro-4-oxopentanoate

  • MDL number MFCD20621661
  • Cas Number : 14594-24-2
  • Molecular Weight : 178.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name ethyl 5-chloro-4-oxopentanoate
Inchi Code 1S/C7H11ClO3/c1-2-11-7(10)4-3-6(9)5-8/h2-5H2,1H3
Inchi Key DPWSGDKZQIYIQC-UHFFFAOYSA-N

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-177417

Safety Information

signalword Danger
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P310,P312,P330,P332+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-177417
pictograms GHS05,GHS07
hstatements H302,H315,H318,H335
Availability details for
Quantity
  • Availability
  • Add to Cart