ENA424046136 Enamine

N-[4-(dimethylamino)phenyl]prop-2-enamide

  • MDL number MFCD12091708
  • Cas Number : 13304-63-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-177429

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-177429
Availability details for
Quantity
  • Availability
  • Add to Cart