ENA424046166 Enamine

5-methyl-1,4-thiazepane

  • MDL number MFCD19159799
  • Cas Number : 7670-50-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-177621

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-177621
Availability details for
Quantity
  • Availability
  • Add to Cart