ENA424046168 Enamine

4-(4-fluorophenoxy)-1-(4-fluorophenyl)-1H-pyrazolo[3,4-d]pyrimidine

  • MDL number MFCD06698417
  • Cas Number : 893966-93-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-177623

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-177623
Availability details for
Quantity
  • Availability
  • Add to Cart