ENA424046171 Enamine

1-(4-chlorophenyl)-5-(propan-2-yl)-1H-pyrazol-4-amine

  • MDL number MFCD21773818
  • Cas Number : 1508227-54-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-177628

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-177628
Availability details for
Quantity
  • Availability
  • Add to Cart