ENA424046172 Enamine

2-(bromomethyl)-2,3-dihydro-1-benzofuran

  • MDL number MFCD00060518
  • Cas Number : 19997-53-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-177630

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-177630
Availability details for
Quantity
  • Availability
  • Add to Cart