ENA424046176 Enamine

ethyl 2-(4-chlorophenyl)-2-(cyclopropylamino)acetate

  • MDL number MFCD12518230
  • Cas Number : 1218642-88-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-177652

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-177652
Availability details for
Quantity
  • Availability
  • Add to Cart