ENA424046180 Enamine

2-(methoxymethylidene)propanedinitrile

  • MDL number MFCD07437817
  • Cas Number : 672-81-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 93-96
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-177658

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-177658
Availability details for
Quantity
  • Availability
  • Add to Cart