ENA424046202 Enamine

1-[5-methyl-1-(pyridin-2-yl)-1H-pyrazol-4-yl]ethan-1-amine

  • MDL number MFCD06809695
  • Cas Number : 956781-21-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-31959

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-31959
Availability details for
Quantity
  • Availability
  • Add to Cart