ENA424046206 Enamine

5,5-bis(butylsulfanyl)pentane-1,2,3,4-tetrol

  • MDL number MFCD28012472

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-43562

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-43562
Availability details for
Quantity
  • Availability
  • Add to Cart