ENA424046218 Enamine

1-cyano-N,N-dimethylmethanesulfonamide

  • MDL number MFCD09041680
  • Cas Number : 926265-18-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-61254

Safety Information

msds https://www.enaminestore.com/catalog/EN300-61254
Availability details for
Quantity
  • Availability
  • Add to Cart