ENA424046221 Enamine

5-fluoro-1-N-propylbenzene-1,2-diamine

  • MDL number MFCD16744197
  • Cas Number : 1072928-93-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-62566

Safety Information

msds https://www.enaminestore.com/catalog/EN300-62566
Availability details for
Quantity
  • Availability
  • Add to Cart