ENA424046224 Enamine

4-chloro-8-fluoroquinoline

  • MDL number MFCD00278785
  • Cas Number : 63010-72-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-67683

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-67683
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart